missing translation for 'onlineSavingsMsg'
Learn More

CHEMGENE MedLab Multi-Surface Disinfectant - Packaging in German, French, Dutch, Portuguese

Product Code. 31451499
Change view
Click to view available options
Color:
Blue
Clear
Scent:
Eucalyptus
Unscented
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Scent
31451499 Clear Unscented
31451498 Blue Eucalyptus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
SAFETY PRECAUTIONS DANGER: Causes skin irritation. Causes serious eye damage. Very toxic to aquatic life with long lasting effects. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wash contaminated skin thoroughly after handling. IF ON SKIN: Wash with plenty of soap and water. Dispose of contents/container in accordance with national regulations.
Product Code. 31451499 Supplier CHEMGENE Supplier No. SKU075B

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy
SAFETY PRECAUTIONS DANGER: Causes skin irritation. Causes serious eye damage. Very toxic to aquatic life with long lasting effects. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wash contaminated skin thoroughly after handling. IF ON SKIN: Wash with plenty of soap and water. Dispose of contents/container in accordance with national regulations.

Fast-acting multi-surface disinfectant for labs, critical services, and forensic settings; decontaminates surfaces and denatures DNA/RNA.

  • CHEMGENE MedLab is a range of surface disinfectants created specifically to meet the demands of laboratory environments
  • These Multi-Surface Disinfectants are proven to be effective under the following microbiological test conditions; Bactericidal: EN 17387, EN 13727 Legionella: EN 13623. Yeasticidal: EN 13624, EN 17387. Fungicidal: EN 17387, EN 13624 Virucidal: EN 14476, EN 16777 (Vaccinia virus, Norovirus, Adenovirus) Bacteriophage: EN 13610
  • Proven efficacy against resistant pathogens
  • Denatures DNA/RNA
  • Biodegradable formulation
  • Available in eucalyptus scent or un-fragranced
  • Manufactured in the UK to ISO 9001 quality standards
  • Non-corrosive in-use
  • Alcohol-free
  • Suitable for autoclaving
  • No rinsing required
  • Compatible with a wide range of materials including mild steel, stainless steel, copper, aluminium, acetal, nylon and rubber.
  • Bottle made from 30% recycled HDPE
  • Bottle and cap recyclable after use

Specifications

Product Type Multi-purpose Cleaner
Color-coded No
Form Concentrated Liquid
pH 10.5 to 11.4
Scent Unscented
Sterility Non-sterile
Autoclavable Autoclavable
Color Clear
Container Type Jerry can - 30% recycled HDPE
Content And Storage Store Out of Direct Sunlight
Dilution 1 to 10%
Dimensions (L x W x H) 133 x 185 x 282 mm
Disposable Limited-use
Filtered Filtered
Flammable No
For Use With (Application) Cleaning and Disinfection of Hard Surfaces and Equipment
Height (Metric) 282 mm
Packaging Bottle - 30% recycled HDPE
Volume (Metric) 5 L
Width (Metric) 185mm
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • X2 MedLab MultiSurface Disinfectant Concentrate 5L Unfragranced
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: N,N-dimethyldecylamine N-oxide, Propan-2-ol, Didecyldimethylammonium chloride, Alcohols, C9-11, etho

Use biocides safely. Always read the product label and information before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.