missing translation for 'onlineSavingsMsg'
Learn More

CHEMGENE MedLab Multi-Surface Disinfectant

Product Code. 30274263 Shop All Chemgene Products
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
30274263 5 L Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30274263 Supplier CHEMGENE Supplier No. SKU075E
Quantity Price
80 £99.00 / Pack of 2
1 £110.00 / Pack of 2
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for a range of disinfection applications within laboratory environments.

  • Next-generation CHEMGENE HLD4L and HLD4H, CHEMGENE MedLab is a range of surface disinfectants created specifically to meet the demands of laboratory environments.
  • CHEMGENE MedLab Multi-Surface Disinfectants are proven to be effective under the following microbiological test conditions; Bactericidal: EN 17387, EN 13727, EN 16615. Legionella: EN 13623. Yeasticidal: EN 13624, EN 17387, EN 16615. Fungicidal: EN 17387, EN 13624. Virucidal: EN 14476, EN 16777 (Norovirus, Adenovirus). Bacteriophage: EN 13610.
  • Broad spectrum protection even in the presence of organic soil
  • Economical dilution rates from 1%
  • Non-corrosive, chlorine-free and alcohol-free
  • Compatible with a wide range of materials including mild steel, stainless steel, copper, aluminium, acetal, nylon and rubber.
  • Complies with relevant CLP, REACH, and biocidal regulations in UK and EU
  • Biodegradable formulation
  • Diluted solution remains effective for 3 months when stored correctly
  • End-user resources available to promote correct use and compliance
  • Manufactured in the UK

Specifications

Product Type Disinfectant Concentrate
Color-coded No
Form Concentrated Liquid
pH 10.5 to 11.4
Scent Unscented
Sterility Non-sterile
Appearance White
Autoclavable Autoclavable
Color White
Container Type Jerry can - UN grade
Content And Storage Store out of direct sunlight. Store at ambient temperature. Store in a closed container. Protect from frost. Protect from heat. Keep away from oxidising substances.
Dilution 1% - 10%
Dimensions (L x W) 134 x 185 mm
Dimensions (L x W x H) 134 x 185 x 284 mm
Filtered Filtered
Flammable No
For Use With (Application) Disinfection of hard surfaces and equipment, cell culture disposal
Height (English) 11.1 in.
Height (Metric) 284 mm
Length (English) 7.2 in.
Length (Metric) 185 mm
Packaging Jerry can - UN grade HDPE
Size 5 L
Volume (English) 1 gal.
Volume (Metric) 5 L
Weight (English) 186 oz.
Weight (Metric) 5282 g
Width (English) 7.2 in.
Width (Metric) 185 mm
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • CHEMGENE MedLab Multi-Surface Disinfectant
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash contaminated skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Didecyldimethylammonium chloride, Alkyl(C12-16) dimethylbenzyl ammonium chloride, N-(3-Aminopropyl)

Use biocides safely. Always read the product label and information before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.