missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Vascular Smooth Muscle Cell Basal Medium (formerly Medium 231)

Product Code. 10781934 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10781934 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10781934 Supplier Gibco™ Supplier No. M231500
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Human Vascular Smooth Muscle Cell Basal Medium formerly Medium 231) is a sterile, liquid culture medium intended for use as one component in a complete culture environment for the growth of normal human vascular smooth muscle cells. When supplemented with SMGS, this medium will support the plating and proliferation of smooth muscle cells at densities between 2.5 x 103 cells/cm2 and 1 x 105 cells/cm2.

  • Additional applications for use may include primary isolation of human smooth muscle cells
  • Medium contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts; does not contain antibiotics, antimycotics, hormones, growth factors, or proteins
  • HEPES and bicarbonate buffered; designed for use in an incubator with an atmosphere of 5% CO2/95% air
  • To support the plating and proliferation of normal human vascular smooth muscle cells, Medium 231 must be supplemented with Smooth Muscle Growth Supplement (SMGS)
  • To induce differentiation of normal human vascular smooth muscle cells, Human Vascular Smooth Muscle Cell Basal Medium must be supplemented with Smooth Muscle Differentiation Supplement (SMDS)
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Smooth Muscle Cells
Form Liquid
Product Type Basal Medium
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives HEPES
Without Additives No Phenol Red
Product Line Cascade Biologics
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.