missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Mammary Epithelial Cell Basal Medium (formerly Medium 171)

Product Code. 10154823 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10154823 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10154823 Supplier Gibco™ Supplier No. M171500
  • ONLINE EXCLUSIVE PRICE
    £88.50
    £78.65 / 500mL
    Save £9.85
    11% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

A sterile, liquid tissue culture media that is intended for use in a complete culture environment for the growth of normal human mammary epithelial cells. Contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts.

Human Mammary Epithelial Cell Basal Medium (formerly 'Medium 171') is a sterile, liquid tissue culture medium intended for use in a complete culture environment for the growth of normal human mammary epithelial cells. This medium contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. It does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. It is HEPES and bicarbonate buffered and designed for use in an incubator with an atmosphere of 5% CO2/95% air.

To support plating and long-term proliferation of normal human mammary epithelial cells, Human Mammary Epithelial Cell Basal Medium must be supplemented with Mammary Epithelial Growth Supplement (MEGS) (Cat. No. S0155). Each 5-mL bottle of MEGS contains all of the growth factors, hormones, and tissue extracts necessary for the culture of normal human mammary epithelial cells and is the correct amount of supplement for a 500 mL bottle of Human Mammary Epithelial Cell Basal Medium.

When a 500 mL bottle of medium is supplemented with MEGS, the final concentrations of the MEGS components in the supplemented medium are: BPE, 0.4% v/v; recombinant human insulin-like growth factor-I, 0.01 μg/mL; hydrocortisone, 0.5 μg/mL; and recombinant human epidermal growth factor, 3 ng/mL.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mammalian Cells
Classification Serum-free
Form Liquid
Product Type Medium 171
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Phenol Red
Product Line Cascade Biologics
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage 4 degrees C
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.