missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Medium 154CF, Kit
GREENER_CHOICE

Product Code. 10144823
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10144823 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10144823 Supplier Gibco™ Supplier No. M154CF500
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Medium 154CF, Kit

A sterile, liquid medium for the culture of human epidermal keratinocytes. Medium 154CF is Medium 154 prepared without calcium chloride. Calcium chloride is provided as a separate component with each bottle of medium. This basal medium requires the addition of HKGS (S-001-5) or HKGS Kit (S-001-K) prior to use.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Keratinocyte Cells
Classification Serum-free
Form Liquid
Product Type Medium 154CF Kit
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Phenol Red
Without Additives No Calcium Chloride
Product Line Cascade Biologics
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage 4 degrees C
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.