missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Medium 131, Kit

Product Code. 10124683
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10124683 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10124683 Supplier Gibco™ Supplier No. M131500

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Medium 131, Kit

A sterile liquid medium for the culture of human microvascular endothelial cells. This basal medium requires the addition of Microvascular Growth Supplement (MVGS) prior to use.

This kit includes one bottle (100 ml) of Attachment Factor.
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Medium 131 Kit
For Use With (Application) Mammalian cell culture, primary cell culture
Quantity 1 Kit
Content And Storage 4°C
Description Medium 131 Kit, 1 Kit, Liquid, Endothelial Cells (Microvascular) Cell Type, Human Species, Phenol Red Indicator, Contains 500mL Medium 131 and 100mL Attachment Factor, For Mammalian Cell Culture, Primary Cell Culture

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.