missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mediator of cell motility 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | mediator of cell motility 1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:100 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232504
|
Novus Biologicals
NBP3-38565-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228151
|
Novus Biologicals
NBP3-38565-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
mediator of cell motility 1 Polyclonal antibody specifically detects mediator of cell motility 1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| mediator of cell motility 1 | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 51072 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C21orf19-like protein, C2orf4DKFZp434I0135, CGI-27, FLJ25031, HCV NS5A-transactivated protein 7, Hepatitis C virus NS5A-transactivated protein 7, mediator of cell motility 1, Mediator of ErbB2-driven cell motility 1, memo-1, MEMOchromosome 2 open reading frame 4, NS5ATP7, protein MEMO1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-297 of human mediator of cell motility 1 (NP_057039.1).,, Sequence:, MSNRVVCREASHAGSWYTASGPQLNAQLEGWLSQVQSTKRPARAIIAPHAGYTYCGSCAAHAYKQVDPSITRRIFILGPSHHVPLSRCALSSVDIYRTPLYDLRIDQKIYGELWKTGMFERMSLQTDEDEHSIEMHLPYTAKAMESHKDEFTIIPVLVGALSESKEQEFGKLFSKYLADPSNLFVVSSDFCHWGQRFRYSYYDESQGEIYRSIEHLDKMGMSIIEQLDPVSFSNYLKKYHNTICGRHPIGVLLNAITELQKNGMNMSFSFLNYAQSSQCRNWQDSSVSYAAGALTVH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title