missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mediator of cell motility 1 Antibody (1E9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22593
This item is not returnable.
View return policy
Description
mediator of cell motility 1 Monoclonal specifically detects mediator of cell motility 1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| mediator of cell motility 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:3000, Flow Cytometry, ELISA 1:100-1:2000 | |
| C21orf19-like protein, C2orf4DKFZp434I0135, CGI-27, FLJ25031, HCV NS5A-transactivated protein 7, Hepatitis C virus NS5A-transactivated protein 7, mediator of cell motility 1, Mediator of ErbB2-driven cell motility 1, memo-1, MEMOchromosome 2 open reading frame 4, NS5ATP7, protein MEMO1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human MEMO1 mAb, clone 1E9, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human MEMO1 protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA | |
| 1000000000 | |
| Unconjugated | |
| NP_057039 | |
| MEMO1 | |
| Recombinant human MEMO1 (1-297aa) purified from E. coli | |
| 0.1 mL | |
| Signal Transduction | |
| 51072 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction