missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEDAG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570956
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 85%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 93%; Rat: 93%.
MEDAG is a protein coding gene involved in processes that promote adipocyte differentiation, lipid accumulation, and glucose uptake in mature adipocytes.
Specifications
| MEDAG | |
| Polyclonal | |
| Unconjugated | |
| MEDAG | |
| 6330406I15Rik; Activated in W/Wv mouse stomach 3; activated in W/Wv mouse stomach 3 homolog; Awms3; C12H13orf33; C13orf33; C25H13orf33; hAWMS3; mAWMS3; MEDA4; MEDA-4; Medag; mesenteric estrogen dependent adipogenesis; mesenteric estrogen-dependent adipogenesis; mesenteric estrogen-dependent adipogenesis protein; Mesenteric estrogen-dependent adipose 4; RGD1307396; uncharacterized protein C13orf33 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84935 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5VYS4 | |
| MEDAG | |
| synthetic peptide directed towards the middle region of human MEDAG. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction