missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528907
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Recommended positive controls: U87-MG. Predicted reactivity: Mouse (96%), Chicken (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein that is one of more than 20 subunits of the mediator complex, first identified in S. cerevisiae, that is required for activation of transcription. The product of this gene also interacts with elongins B and C, and CUL2 and RBX1, to reconstitute a ubiquitin ligase. Two alternative transcripts encoding different isoforms have been described.
Spécification
| MED8 | |
| Polyclonal | |
| Unconjugated | |
| Med8 | |
| 2210021A15Rik; AB041805; Activator-recruited cofactor 32 kDa component; ARC32; Med8; Mediator complex subunit 8; mediator of RNA polymerase II transcription subunit 8; mediator of RNA polymerase II transcription subunit MED8; mediator of RNA polymerase II transcription, subunit 8 homolog; MGC17544; MGC19641; MNCb-2386; zgc:76877; zgc:76877 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 112950 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96G25 | |
| Med8 | |
| Recombinant protein encompassing a sequence within the center region of human MED8. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu