missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-98339
This item is not returnable.
View return policy
Description
MED7 Polyclonal specifically detects MED7 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Chromatin Immunoprecipitation (ChIP).
Specifications
| MED7 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Activator-recruited cofactor 34 kDa component, Cofactor required for Sp1 transcriptional activation subunit 9, cofactor required for Sp1 transcriptional activation, subunit 9 (33kD), cofactor required for Sp1 transcriptional activation, subunit 9, 33kDa, CRSP33CRSP complex subunit 9, CRSP9hMED7, mediator complex subunit 7ARC34, mediator of RNA polymerase II transcription subunit 7, MGC12284, RNA polymerase transcriptional regulation mediator subunit 7 homolog, Transcriptional coactivator CRSP33 | |
| Rabbit | |
| 27 kDa | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 9443 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot, ChIP Assay | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| NP_004261 | |
| MED7 | |
| The immunogen for this antibody is MED7 - C-terminal region. Peptide sequence LASLPDDLPHSEAGMRVKTEPMDADDSNNCTGQNEHQRENSGHRRDQIIE. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction