missing translation for 'onlineSavingsMsg'
Learn More

MED6 Antibody (4C2), Novus Biologicals™

Product Code. 18353739 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353739 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353739 Supplier Novus Biologicals Supplier No. H00010001M07

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MED6 Monoclonal antibody specifically detects MED6 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED6
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4C2
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH04106
Gene Alias Activator-recruited cofactor 33 kDa component, ARC33, mediator complex subunit 6hMed6, mediator of RNA polymerase II transcription subunit 6, mediator of RNA polymerase II transcription, subunit 6 homolog, mediator of RNA polymerase II transcription, subunit 6 homolog (S. cerevisiae), NY-REN-28, Renal carcinoma antigen NY-REN-28
Host Species Mouse
Immunogen MED6 (AAH04106, 1 a.a. ~ 246 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAVDIRDNLLGISWVDSSWIPILNSGSVLDYFSERSNPFYDRTCNNEVVKMQRLTLEHLNQMVGIEYILLHAQEPILFIIRKQQRQSPAQVIPLADYYIIAGVIYQAPDLGSVINSRVLTAVHGIQSAFDEAMSYCRYHPSKGYWWHFKDHEEQDKVRPKAKRKEEPSSIFQRQRVDALLLDLRQKFPPKFVQLKPGEKPVPVDQTKKEAEPIPETVKPEEKETTKNVQQTVSAKGPPEKRMRLQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10001
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.