Learn More
Invitrogen™ MED4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597188
Description
Immunogen sequence: MAASSSGEKE KERLGGGLGV AGGNSTRERL LSALEDLEVL SRELIEMLAI SRNQKLLQAG EENQVLELLI HRDGEFQELM KLALNQGKIH HEMQVLEKEV EKRDSDIQQL QKQLKEAEQI LATAVYQAKE KLKSIEKARK GAISSEEIIK YAHRISASNA VCAPLTWVPG DPRRPYPTDL EMRSGLLGQM NNPSTNGVNG HLPGDALAAG RLPDVLAPQY PWQSNDMSMN MLPPNHSSDF LLEPPGHNKE NEDDVEIMST DSSSSSSESD; Positive Samples: Jurkat, SH-SY5Y, HL-60, SW480, HT-1080; Cellular Location: Nucleus.
The Mediator is a multiprotein coactivator that is required by DNA-binding transcription factors for activation of polymerase II transcribed genes. MED4 appears to be a core Mediator subunit and is found in nearly all Mediator preparations (Sato et al., 2004).
Specifications
| MED4 | |
| Polyclonal | |
| Unconjugated | |
| MED4 | |
| 2410046H15Rik; Activator-recruited cofactor 36 kDa component; ARC36; DRIP36; FLJ10956; HSPC126; hypothetical protein LOC550582; LOW QUALITY PROTEIN: mediator of RNA polymerase II transcription subunit 4; MED4; Mediator complex subunit 4; mediator of RNA polymerase II transcription subunit 4; mediator of RNA polymerase II transcription, subunit 4 homolog; mediator, 34-kD subunit, homolog; p36 TRAP/SMCC/PC2 subunit; RP11-90M2.2; TRAP/SMCC/PC2 subunit p36; TRAP/SMCC/PC2 subunit p36 subunit; TRAP36; Vdirp; Vdrip; vitamin D receptor interacting protein; vitamin D receptor-interacting protein, 36-kD; vitamin D3 receptor-interacting protein complex 36 kDa component; zgc:110646 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 29079 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.03 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NPJ6 | |
| MED4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human MED4 (NP_0548851). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.