missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76930
This item is not returnable.
View return policy
Description
MED4 Polyclonal specifically detects MED4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MED4 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Activator-recruited cofactor 36 kDa component, DRIP36mediator of RNA polymerase II transcription, subunit 4 homolog (S. cerevisiae), FLJ10956, HSPC126vitamin D receptor interacting protein, mediator complex subunit 4TRAP/SMCC/PC2 subunit p36 subunit, subunit 4 homolog, TRAP36 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| At least three isoforms of MED4 are known to exist; this antibody only recognizes the largest isoform. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
| NP_054885 | |
| MED4 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human MED4. The immunogen is located within the last 50 amino acids of MED4. Amino Acid Squence: PGHNKEDEDDVEIMSTD | |
| 0.1 mg | |
| Signal Transduction | |
| 29079 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction