missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115564
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MED31.
MED31 is the component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes.
Specifications
| MED31 | |
| Polyclonal | |
| Unconjugated | |
| MED31 | |
| 3110004H13Rik; CGI-125; fa04d01; FLJ27436; FLJ36714; hSOH1; l11Jus15; med31; Mediator complex subunit 31; mediator complex subunit soh1; Mediator of RNA polymerase II transcription subunit 31; mediator of RNA polymerase II transcription subunit 31; LOW QUALITY PROTEIN: mediator of RNA polymerase II transcription subunit 31; mediator of RNA polymerase II transcription, subunit 31 homolog; RGD1309457; soh1; wu:fa04d01; zgc:92673 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 287475, 51003, 67279 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9CXU1, Q9Y3C7 | |
| MED31 | |
| A synthesized peptide derived from human MED31(Accession Q9Y3C7), corresponding to amino acid residues M1-N35. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction