missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED30 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16787-1-AP-20UL
This item is not returnable.
View return policy
Description
Med30 is a subunit of the RNA polymerase II (Pol II) transcriptional mediator complex. The mediator complex is a coactivator involved in the regulated transcription of Pol II-dependent genes. The mediator complex functions as a bridge to convey information from gene-specific regulatory proteins to the basal Pol II transcription machinery, and is recruited to promoter regions by directly interacting with regulatory proteins. The mediator complex also serves as a scaffold for the assembly of a functional pre-initiation complex with Pol II and other general transcription factors. Found in the nucleus, Med30 is expressed in brain, heart, kidney, liver, lung, pancreas, placenta and skeletal muscle.Specifications
| MED30 | |
| Polyclonal | |
| Unconjugated | |
| MED30 | |
| MED30, mediator complex subunit 30, THRAP6, TRAP25 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 69790, 90390 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96HR3, Q9CQI9 | |
| Med30 | |
| MED30 Fusion Protein Ag10221 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction