Learn More
MED30 Mouse anti-Human, Unconjugated, Clone: 1D7C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67038-1-IG-150UL
This item is not returnable.
View return policy
Description
The MED30 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MED30. This antibody recognizes human, rat, pig antigen. The MED30 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| MED30 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Pig |
| Western Blot, ELISA | |
| 1D7C4 | |
| Unconjugated | |
| 150 μL | |
| 90390 | |
| IgG2a |