missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED26 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567203
This item is not returnable.
View return policy
Description
Immunogen sequence: ALDATQVPSPL PLAQPSTPPV RRLELLPSAE SPVCWLEQPE SHQRLAGPGC KAGLSPAEPL LSRAGFSPDS S Highest antigen sequence identity to the following orthologs - mouse 58%, rat 63%.
This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.
Specifications
| MED26 | |
| Polyclonal | |
| Unconjugated | |
| MED26 | |
| 5730493L18Rik; Activator-recruited cofactor 70 kDa component; AI414941; ARC70; AW495270; cofactor required for Sp1 transcriptional activation subunit 7; cofactor required for Sp1 transcriptional activation, subunit 7 (70kD); cofactor required for Sp1 transcriptional activation, subunit 7, 70kDa; CRSP complex subunit 7; Crsp7; CRSP70; Med26; Mediator complex subunit 26; mediator of RNA polymerase II transcription subunit 26; Slc35e1; solute carrier family 35, member E1; transcriptional coactivator CRSP70 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9441 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95402 | |
| MED26 | |
| Recombinant Human MED26. Recombinant protein control fragment (Product #RP-107839). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction