missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MED25 Monoclonal Antibody (7F1)
GREENER_CHOICE

Product Code. 15672295
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15672295 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15672295 Supplier Invitrogen™ Supplier No. MA524469

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: MASAGVAAGR QAEDVLPPTS DQPLPDTKPL PPPQPPPVPA PQPQQSPAPR PQSPARAREE ENYSFLPLVH NIIKCMDKDS PEVHQDLNAL KSKFQEMRKL ISTMPGIHLS PEQQQQQLQS LREQVRTKNE LLQKYKSLCM FEIPKE.

This gene encodes a component of the transcriptional coactivator complex termed the Mediator complex. This complex is required for transcription of most RNA polymerase II-dependent genes. The encoded protein plays a role in chromatin modification and in preinitiation complex assembly. Mutations in this gene are associated with Charcot-Marie-Tooth disease type 2B2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED25
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7F1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene MED25
Gene Accession No. Q71SY5
Gene Alias 2610034E13Rik; 2610529E18Rik; ACID1; Activator interaction domain-containing protein 1; activator-recruited cofactor 92 kDa component; ARC/mediator transcriptional coactivator subunit; ARC92; BVSYS; CMT2B2; DKFZp434K0512; ESTM2; hypothetical protein LOC406793; LOW QUALITY PROTEIN: mediator of RNA polymerase II transcription subunit 25; MED25; Mediator complex subunit 25; mediator of RNA polymerase II transcription subunit 25; mediator of RNA polymerase II transcription, subunit 25 homolog; mediator of RNA polymerase II transcription, subunit 9 homolog; MGC70671; mMED25; P78; prostate-derived protein 78 kDa; PTOV2; RGD1305137; TCBAP0758; zgc:63555
Gene Symbols MED25
Host Species Mouse
Immunogen MED9 (NP_060489.1, 1 a.a. ~ 146 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81857
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.