missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED25 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £484.00
Specifications
| Antigen | MED25 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18277554
|
Novus Biologicals
NBP2-55868 |
100 μL |
£484.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18697218
|
Novus Biologicals
NBP2-55868-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MED25 Polyclonal specifically detects MED25 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| MED25 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 81857 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PPGAASGPPPPGPILRPQNPGANPQLRSLLLNPPPPQTGVPPPQASLHHLQPPG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| ACID1, Activator Interaction Domain-Containing Protein 1, Activator-Recruited Cofactor 92 KDa Component, ARC/Mediator Transcriptional Coactivator Subunit, ARC92, BVSYS, CMT2B2, Mediator Complex Subunit 25, Mediator Of RNA Polymerase II Transcription Subunit 25, Mediator Of RNA Polymerase II Transcription, Subunit 25 Homolog, Mediator Of RNA Polymerase II Transcription, Subunit 25 Homolog (S. Cerevisiae), P78, TCBAP0758 | |
| MED25 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title