missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED24 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74599AF700
This item is not returnable.
View return policy
Description
MED24 Polyclonal antibody specifically detects MED24 in Human, Mouse samples. It is validated for Western Blot
Specifications
| MED24 | |
| Polyclonal | |
| Western Blot | |
| cofactor required for Sp1 transcriptional activation, subunit 4, 100kDa, CRSP100, DRIP100CRSP complex subunit 4, hTRAP100, KIAA0130CRSP4, mediator complex subunit 24ARC100, MGC8748, subunit 24 homolog, thyroid hormone receptor associated protein 4, Thyroid hormone receptor-associated protein 4, Thyroid hormone receptor-associated protein complex 100 kDa component, Trap100, TRAP100THRAP4, Vitamin D3 receptor-interacting protein complex 100 kDa component, vitamin D3 receptor-interacting protein complex component DRIP100 | |
| The immunogen recognized by this antibody maps to a region between residue 939 and 989 of human thyroid hormone receptor-associated protein complex 100 kDa component (mediator complex subunit 24 ) using the numbering given in entry NP_055630.2 (GeneID 986). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9862 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction