missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED23 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27850-1-AP-20UL
This item is not returnable.
View return policy
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. DDX50 is a DEAD box enzyme that may be involved in ribosomal RNA synthesis or processing. DDX50 and DDX21, also called RH-II/GuA, have similar genomic structures and are in tandem orientation on chromosome 10, suggesting that the two genes arose by gene duplication in evolution. DDX50 gene has pseudogenes on chromosomes 2, 3 and 4. Alternative splicing of this gene generates multiple transcript variants, but the full length nature of all the other variants but one has not been defined.Specifications
| MED23 | |
| Polyclonal | |
| Unconjugated | |
| MED23 | |
| ARC130, CRSP complex subunit 3, CRSP130, CRSP133, CRSP3, DRIP130, hSur 2, KIAA1216, MED23, mediator complex subunit 23, Protein sur 2 homolog, SUR2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9439 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9ULK4 | |
| MED23 | |
| MED23 Fusion Protein Ag27241 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction