missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED23 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB200-337
This item is not returnable.
View return policy
Description
MED23 Polyclonal specifically detects MED23 in Human, Mouse samples. It is validated for Chromatin Immunoprecipitation, Immunoprecipitation, Western Blot (Negative).
Specifications
| MED23 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| 130 kDa transcriptional co-activator, 133 kDa transcriptional co-activator, CRSP130, DRIP130DKFZp434H0117, mediator complex subunit 23CRSP133, Protein sur-2 homolog, subunit 3, 130kDa, Transcriptional coactivator CRSP130, Vitamin D3 receptor-interacting protein complex 130 kDa component | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB200-337 is specific for human CRSP 3 protein. | |
| Store at 4C. Do not freeze. |
| Immunoprecipitation, Western Blot, ChIP Assay | |
| 1.0 mg/mL | |
| Immunoprecipitation 1-4 ug/mg lysate, Western Blot (Negative), Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Q9ULK4 | |
| MED23 | |
| A synthetic peptide that maps to a region between residues 1 and 50 of human Cofactor Required for Sp1 transcriptional activation, subunit 3 using the numbering given in Swiss-Prot entry Q9ULK4 (GeneID 9439). | |
| 100 μL | |
| Cell Biology, Signal Transduction, Transcription Factors and Regulators | |
| 9439 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction