missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103916
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MED22.
This gene is located in the surfeit gene cluster, a group of very tightly linked housekeeping genes that do not share sequence similarity. The gene is oriented in a head-to-head fashion with RPL7A (SURF3) and the two genes share a bidirectional promoter. The encoded proteins are localized to the cytoplasm. Two alternative transcript variants encoding different isoforms have been identified for this gene.
Specifications
| MED22 | |
| Polyclonal | |
| Unconjugated | |
| MED22 | |
| AW212655; AW558812; MED22; MED24; Mediator complex subunit 22; mediator of RNA polymerase II transcription subunit 22; MGC48682; RGD1564893; si:ch73-278c20.1; si:dkey-226m8.2; surf5; Surf-5; surfeit 5; surfeit 5 isoform b; surfeit gene 5; surfeit locus protein 5; wu:fj45h07; zgc:86721 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20933, 499762, 6837 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A0JPN6, Q15528, Q62276 | |
| MED22 | |
| A synthesized peptide derived from human MED22(Accession Q15528), corresponding to amino acid residues L119-P169. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction