missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED21 Antibody (PCRP-MED21-4B5) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14069JF585
This item is not returnable.
View return policy
Description
MED21 Monoclonal antibody specifically detects MED21 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Western Blot,Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| MED21 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| hSrb7, mediator complex subunit 21SRB7 (suppressor of RNA polymerase B, yeast) homolog, RNA polymerase II complex component SRB7, RNA polymerase II holoenzyme component SRB7, RNAPII complex component SRB7, SRB7 suppressor of RNA polymerase B homolog, SRB7SRB7 suppressor of RNA polymerase B homolog (yeast), SURB7mediator of RNA polymerase II transcription subunit 21 | |
| Recombinant full-length human MED21 protein (Uniprot: Q13503) | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 9412 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunoprecipitation, Western Blot, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-MED21-4B5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction