missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | MED20 |
|---|---|
| Concentration | 0.18 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16837534
|
Proteintech
17598-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16827534
|
Proteintech
17598-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MED20 is a subunit of the Mediator complex, a multiprotein coactivator of RNA transcription that interacts with DNA-bound transcriptional activators, RNA polymerase II, and general initiation factors.Specifications
| MED20 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| Q5XIE9, Q9H944, Q9R0X0 | |
| 316209, 56771, 9477 | |
| MED20 Fusion Protein Ag11598 | |
| Primary | |
| -20°C | |
| MED20 |
| 0.18 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| hTRFP, MED20, mediator complex subunit 20, PRO0213, TRF proximal protein homolog, TRFP | |
| Med20 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title