missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100799
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MED20.
MED20 is a subunit of the Mediator complex, a multiprotein coactivator of RNA transcription that interacts with DNA-bound transcriptional activators, RNA polymerase II, and general initiation factors.
Specifications
| MED20 | |
| Polyclonal | |
| Unconjugated | |
| MED20 | |
| 1110011O05Rik; 2410115I17Rik; AU018348; hTRFP; Med20; Mediator complex subunit 20; mediator of RNA polymerase II transcription subunit 20; PRO0213; Trf (TATA binding protein-related factor)-proximal homolog; Trf (TATA binding protein-related factor)-proximal protein homolog; TRFP; TRF-proximal protein homolog; ubiquitin specific protease 49; Usp49 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 316209, 56771, 9477 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5XIE9, Q9H944, Q9R0X0 | |
| MED20 | |
| A synthesized peptide derived from human MED20(Accession Q9H944), corresponding to amino acid residues K27-A77. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction