Learn More
Invitrogen™ MED20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590677
Description
Immunogen sequence: MGVTCVSQMP VAEGKSVQQT VELLTRKLEM LGAEKQGTFC VDCETYHTAA STLGSQGQTG KLMYVMHNSE YPLSCFALFE NGPCLIADTN FDVLMVKLKG FFQSAKASKI ETRGTRYQYC DFLVKVGTVT MGPSARGISV EVEYGPCVVA SDCWSLLLEF LQSFLGSHTP GAPAVFGNRH DAVYGPADTM VQYMELFNKI RKQQQVPVAG IR; Positive Samples: DU145, Mouse brain; Cellular Location: Nucleus.
MED20 is a subunit of the Mediator complex, a multiprotein coactivator of RNA transcription that interacts with DNA-bound transcriptional activators, RNA polymerase II, and general initiation factors.
Specifications
| MED20 | |
| Polyclonal | |
| Unconjugated | |
| MED20 | |
| 1110011O05Rik; 2410115I17Rik; AU018348; hTRFP; Med20; Mediator complex subunit 20; mediator of RNA polymerase II transcription subunit 20; PRO0213; Trf (TATA binding protein-related factor)-proximal homolog; Trf (TATA binding protein-related factor)-proximal protein homolog; TRFP; TRF-proximal protein homolog; ubiquitin specific protease 49; Usp49 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 316209, 56771, 9477 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.32 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5XIE9, Q9H944, Q9R0X0 | |
| MED20 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-212 of human MED20 (NP_0042662). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.