missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15352-1-AP-150UL
This item is not returnable.
View return policy
Description
Zinc, an essential element required for cell proliferation and differentiation, plays a role in a diverse array of cellular functions, including acting as a cofactor for numerous enzymes and transcription factors and as a neuroregulator. The zinc transporter (ZnT) family regulates the supply of zinc within cells, and its members are characterized by containing six membrane-spanning domains, a large histidine-rich intracellular loop, and a C-terminal tail. ZnT-1, a ubiquitous protein of approximately 60 kDa, localizes to the plasma membrane to aid in the export of zinc out of cells.Specifications
| MED18 | |
| Polyclonal | |
| Unconjugated | |
| MED18 | |
| MED18, mediator complex subunit 18, p28b | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54797, 67219, 682988 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BUE0, Q9CZ82 | |
| MED18 | |
| MED18 Fusion Protein Ag7598 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction