Learn More
Invitrogen™ MED18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120283
Description
Positive test controls include: HT-29, Jurkat, Mouse liver, Mouse thymus. The target is usually found in the following locations: Nucleus. Immunogen sequence: MEAPPVTMMP VTGGTINMME YLLQGSVLDH SLESLIHRLR GLCDNMEPET FLDHEMVFLL KGQQASPFVL RARRSMDRAG APWHLRYLGQ PEMGDKNRHA LVRNCVDIAT SENLTDFLME MGFRMDHEFV AKGHLFRKGI MKIMVYKIFR ILVPGNTDST EALSLSYLVE LSVVAPAGQD MVSDDMKNFA EQLKPLVHLE KIDPKRLM.
Zinc, an essential element required for cell proliferation and differentiation, plays a role in a diverse array of cellular functions, including acting as a cofactor for numerous enzymes and transcription factors and as a neuroregulator. The zinc transporter (ZnT) family regulates the supply of zinc within cells, and its members are characterized by containing six membrane-spanning domains, a large histidine-rich intracellular loop, and a C-terminal tail. ZnT-1, a ubiquitous protein of approximately 60 kDa, localizes to the plasma membrane to aid in the export of zinc out of cells.
Specifications
| MED18 | |
| Polyclonal | |
| Unconjugated | |
| MED18 | |
| 2810046C01Rik; FLJ20045; Med18; mediator 18; Mediator complex subunit 18; mediator of RNA polymerase II transcription subunit 18; mediator of RNA polymerase II transcription, subunit 18 homolog; p28b; TRAP/mediator complex subunit p28b | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54797, 67219 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.47 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9BUE0, Q9CZ82 | |
| MED18 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-208 of human MED18 (NP_001120822.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.