missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED17 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | MED17 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18333215
|
Novus Biologicals
NBP3-04651-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18331997
|
Novus Biologicals
NBP3-04651-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MED17 Polyclonal specifically detects MED17 in Human samples. It is validated for Western Blot.Specifications
| MED17 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Activator-recruited cofactor 77 kDa component, Cofactor required for Sp1 transcriptional activation subunit 6, cofactor required for Sp1 transcriptional activation, subunit 6, 77kDa, CRSP6FLJ10812, CRSP77cofactor required for Sp1 transcriptional activation, subunit 6 (77kD), DRIP77, DRIP80ARC77, mediator complex subunit 17Vitamin D3 receptor-interacting protein complex 80 kDa component, mediator of RNA polymerase II transcription subunit 17, Thyroid hormone receptor-associated protein complex 80 kDa component, Transcriptional coactivator CRSP77, Trap80, TRAP80CRSP complex subunit 6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human MED17 (NP_004259.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 9440 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title