missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED17 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-17737-100UL
This item is not returnable.
View return policy
Description
MED17 Polyclonal antibody specifically detects MED17 in Human samples. It is validated for Immunofluorescence
Specifications
| MED17 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Activator-recruited cofactor 77 kDa component, Cofactor required for Sp1 transcriptional activation subunit 6, cofactor required for Sp1 transcriptional activation, subunit 6, 77kDa, CRSP6FLJ10812, CRSP77cofactor required for Sp1 transcriptional activation, subunit 6 (77kD), DRIP77, DRIP80ARC77, mediator complex subunit 17Vitamin D3 receptor-interacting protein complex 80 kDa component, mediator of RNA polymerase II transcription subunit 17, Thyroid hormone receptor-associated protein complex 80 kDa component, Transcriptional coactivator CRSP77, Trap80, TRAP80CRSP complex subunit 6 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: IGVEQIRVVHRDGRVITLSYQEQELQDFLLSQMSQHQVHAVQQLAKVMGWQVLSFSNHVGLGPIESIGNASAITVASPSGDYAISVRNGPESGSKIMVQFPRNQCKDLPKSDVLQDNKWSHLRGPFKEVQWNKMEGRN | |
| 100 μg | |
| Signal Transduction | |
| 9440 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction