missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11505-1-AP-20UL
This item is not returnable.
View return policy
Description
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by CRSP6 is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e. g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors.Specifications
| MED17 | |
| Polyclonal | |
| Unconjugated | |
| MED17 | |
| ARC77, CRSP complex subunit 6, CRSP6, CRSP77, DRIP77, DRIP80, MED17, mediator complex subunit 17, TRAP80 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 234959, 300367, 9440 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8VCD5, Q9NVC6 | |
| MED17 | |
| MED17 Fusion Protein Ag2057 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction