missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115563
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MED17.
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by CRSP6 is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e. g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors.
Specifications
| MED17 | |
| Polyclonal | |
| Unconjugated | |
| MED17 | |
| 77kDa; activator-recruited cofactor 77 kDa component; ARC77; C330002H14Rik; cofactor required for Sp1 transcriptional activation subunit 6; cofactor required for Sp1 transcriptional activation, subunit 6; cofactor required for Sp1 transcriptional activation, subunit 6 (77kD); cofactor required for Sp1 transcriptional activation, subunit 6, 77kDa; CRSP complex subunit 6; CRSP6; CRSP77; DRIP77; DRIP80; Med17; Mediator complex subunit 17; mediator of RNA polymerase II transcription subunit 17; Thyroid hormone receptor-associated protein complex 80 kDa component; transcriptional coactivator CRSP77; Trap80; Vitamin D3 receptor-interacting protein complex 80 kDa component | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 234959, 9440 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8VCD5, Q9NVC6 | |
| MED17 | |
| A synthesized peptide derived from human MED17(Accession Q9NVC6), corresponding to amino acid residues E186-I236. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction