missing translation for 'onlineSavingsMsg'
Learn More

MED17 Antibody (1B5), Novus Biologicals™

Product Code. 18327258 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327258 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327258 Supplier Novus Biologicals Supplier No. H00009440M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MED17 Monoclonal antibody specifically detects MED17 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED17
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1B5
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH21101
Gene Alias Activator-recruited cofactor 77 kDa component, Cofactor required for Sp1 transcriptional activation subunit 6, cofactor required for Sp1 transcriptional activation, subunit 6, 77kDa, CRSP6FLJ10812, CRSP77cofactor required for Sp1 transcriptional activation, subunit 6 (77kD), DRIP77, DRIP80ARC77, mediator complex subunit 17Vitamin D3 receptor-interacting protein complex 80 kDa component, mediator of RNA polymerase II transcription subunit 17, Thyroid hormone receptor-associated protein complex 80 kDa component, Transcriptional coactivator CRSP77, Trap80, TRAP80CRSP complex subunit 6
Host Species Mouse
Immunogen CRSP6 (AAH21101, 551 a.a. ~ 651 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. FSNHVGLGPIESIGNASAITVASPSGDYAISVRNGPESGSKIMVQFPRNQCKDLPKSDVLQDNKWSHLRGPFKEVQWNKMEGRNFVYKMELLMSALSPCLL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9440
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.