missing translation for 'onlineSavingsMsg'
Learn More

MED16 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18428647 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428647 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428647 Supplier Novus Biologicals Supplier No. NBP204141AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MED16 Polyclonal antibody specifically detects MED16 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED16
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DRIP92thyroid hormone receptor associated protein 5, mediator complex subunit 16THRAP5, mediator of RNA polymerase II transcription subunit 16, Thyroid hormone receptor-associated protein 5, Thyroid hormone receptor-associated protein complex 95 kDa component, thyroid hormone receptor-associated protein, 95-kD subunit, TRAP95, Vitamin D3 receptor-interacting protein complex 92 kDa component
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 550 and 600 of human Mediator Complex Subunit 16 using the numbering given in entry NP_005472.2 (GeneID 10025).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10025
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.