missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED15 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11566-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a transcription factor that represses GATA-regulated genes and binds to a dynein light chain protein. Binding of the encoded protein to the dynein light chain protein affects binding to GATA consensus sequences and suppresses its transcriptional activity. Defects in this gene are a cause of tricho-rhino-phalangeal syndrome (TRPS) types I-III.Specifications
| MED15 | |
| Polyclonal | |
| Unconjugated | |
| MED15 | |
| ARC105, CAG7A, CTG repeat protein 7a, CTG7A, MED15, mediator complex subunit 15, PCQAP, TIG 1, TIG1, TNRC7, TPA inducible gene 1 protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51586 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96RN5 | |
| MED15 | |
| MED15 Fusion Protein Ag2143 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction