missing translation for 'onlineSavingsMsg'
Learn More

MED15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18377452 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377452 20 μg 20µL
18347184 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18377452 Supplier Novus Biologicals Supplier No. NBP31556520UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MED15 Polyclonal antibody specifically detects MED15 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED15
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias ARC105Arc105, CAG7A, CTG repeat protein 7a, CTG7A, FLJ42282, FLJ42935, mediator complex subunit 15PC2 glutamine/Q-rich-associated protein, mediator of RNA polymerase II transcription subunit 15, PC2 (positive cofactor 2, multiprotein complex) glutamine/Q-rich-associatedprotein, PC2-glutamine-rich-associated protein, PCQAPDKFZp686A2214, Positive cofactor 2 glutamine/Q-rich-associated protein, positive cofactor 2, glutamine/Q-rich-associated protein, TIG1DKFZp762B1216, TIG-1TPA-inducible gene 1 protein, TNRC7, TPA inducible gene-1, TPA inducible protein, trinucleotide repeat containing 7,105-kD, Trinucleotide repeat-containing gene 7 protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MED15 (NP_001003891.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51586
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.