missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED13L Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558789
This item is not returnable.
View return policy
Description
Immunogen sequence: PALYGNGLEL QQLSTLDDRT VLVGQRLPLM AEVSETALYC GIRPSNPESS EKWWHSYRLP PSDDAEFRPP ELQGERCDAK MEVNSESTAL QRLLAQPNKR FKIWQD Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.
The protein encoded by this gene is a subunit of the Mediator complex, a large complex of proteins that functions as a transcriptional coactivator for most RNA polymerase II-transcribed genes. The encoded protein is involved in early development of the heart and brain. Defects in this gene are a cause of transposition of the great arteries, dextro-looped (DTGA).[provided by RefSeq, Jul 2010].
Specifications
| MED13L | |
| Polyclonal | |
| Unconjugated | |
| MED13L | |
| 2210413I17Rik; 6330591G05Rik; 9030618F05Rik; Kiaa1025; Med13l; mediator complex subunit 13 like; mediator complex subunit 13-like; Mediator of RNA polymerase II transcription subunit 13-like; mKIAA1025; MRFACD; PROSIT240; Thrap2; thyroid hormone receptor associated protein 2; thyroid hormone receptor-associated protein 2; Thyroid hormone receptor-associated protein complex 240 kDa component-like; TRAP240L | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23389 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q71F56 | |
| MED13L | |
| Recombinant protein corresponding to Human MED13L. Recombinant protein control fragment (Product #RP-97716). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction