missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535924
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (94%), Rat (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a component of the mediator complex (also known as TRAP, SMCC, DRIP, or ARC), a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. The product of this gene is proposed to form a sub-complex with MED12, cyclin C, and CDK8 that can negatively regulate transactivation by mediator. [provided by RefSeq].
Specifications
| MED13 | |
| Polyclonal | |
| Unconjugated | |
| MED13 | |
| 1110067M05Rik; activator-recruited cofactor 250 kDa component; ARC250; D030023K18; DRIP250; HSPC221; Kiaa0593; Med13; Mediator complex subunit 13; mediator of RNA polymerase II transcription subunit 13; mediator of RNA polymerase II transcription, subunit 13; mediator of RNA polymerase II transcription, subunit 13 homolog; Thrap1; thyroid hormone receptor associated protein 1; thyroid hormone receptor associated protein, 240 kDa subunit; thyroid hormone receptor-associated protein 1; Thyroid hormone receptor-associated protein complex 240 kDa component; thyroid hormone receptor-associated protein complex component TRAP240; thyroid hormone receptor-associated protein, 240 kDa subunit; Trap240; Vitamin D3 receptor-interacting protein complex component DRIP250; Zmediator complex subunit 13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 327987, 9969 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q5SWW4, Q9UHV7 | |
| MED13 | |
| Recombinant protein encompassing a sequence within the center region of human MED13. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction