missing translation for 'onlineSavingsMsg'
Learn More

MED13 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£328.00 - £513.00

Specifications

Antigen MED13
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18243403
View Documents
Novus Biologicals
NBP2-57545
100 μL
£513.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18673369
View Documents
Novus Biologicals
NBP2-57545-25ul
25 μL
£328.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MED13 Polyclonal specifically detects MED13 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

MED13
Polyclonal
Rabbit
Human
Activator-recruited cofactor 250 kDa component, KIAA0593HSPC221, mediator complex subunit 13ARC250, mediator of RNA polymerase II transcription, subunit 13 homolog, THRAP1mediator of RNA polymerase II transcription subunit 13, thyroid hormone receptor associated protein 1, Thyroid hormone receptor-associated protein 1, Thyroid hormone receptor-associated protein complex 240 kDa component, thyroid hormone receptor-associated protein complex component TRAP240, thyroid hormone receptor-associated protein, 240 kDa subunit, Trap240, TRAP240DRIP250, Vitamin D3 receptor-interacting protein complex component DRIP250
MED13
IgG
Affinity Purified
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
9969
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SASVQVASATYTTENLDLAFNPNNDGADGMGIFDLLDTGDDLDPDIINILPASPTGSPVHSPGSHYPHGGDAGKGQSTDRLLSTEPH
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.