missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60641
This item is not returnable.
View return policy
Description
MED13 Polyclonal specifically detects MED13 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| MED13 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| Activator-recruited cofactor 250 kDa component, KIAA0593HSPC221, mediator complex subunit 13ARC250, mediator of RNA polymerase II transcription, subunit 13 homolog, THRAP1mediator of RNA polymerase II transcription subunit 13, thyroid hormone receptor associated protein 1, Thyroid hormone receptor-associated protein 1, Thyroid hormone receptor-associated protein complex 240 kDa component, thyroid hormone receptor-associated protein complex component TRAP240, thyroid hormone receptor-associated protein, 240 kDa subunit, Trap240, TRAP240DRIP250, Vitamin D3 receptor-interacting protein complex component DRIP250 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9969 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500 | |
| NP_005112.2 | |
| MED13 | |
| The immunogen recognized by this antibody maps to a region between residue 325 and 375 of human thyroid hormone receptor associated protein 1 (thyroid hormone receptor-associated protein, 240 kDa) using the numbering given in entry NP_005112.2 (GeneID 996 | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction