missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED12 Recombinant Rabbit Monoclonal Antibody (BLR084G)

Rabbit Recombinant Monoclonal Antibody
£128.00 - £385.00
Specifications
| Antigen | MED12 |
|---|---|
| Clone | BLR084G |
| Concentration | 0.25 mg/mL |
| Content And Storage | 4°C |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17755729
![]() |
Invitrogen™
MA544359 |
100 μL |
£385.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17765729
|
Invitrogen™
MA544360 |
10 μL |
£128.00
10µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
The initiation of transcription is controlled in part by a large protein assembly known as the preinitiation complex. A component of this preinitiation complex is a 1.2 MDa protein aggregate called Mediator. This Mediator component binds with a CDK8 subcomplex which contains the protein encoded by this gene, mediator complex subunit 12 (MED12), along with MED13, CDK8 kinase, and cyclin C.Specifications
| MED12 | |
| 0.25 mg/mL | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
| Q93074 | |
| 9968 | |
| A peptide representing a region between residue 2127 and the C-terminus (residue 2177) of human mediator complex subunit 12. | |
| Antibody |
| BLR084G | |
| 4°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Med12 | |
| 230kDa; activator-recruited cofactor 240 kDa component; ARC240; CAG repeat protein 45; CAGH45; FGS1; HOPA; human opposite paired; Kiaa0192; Med12; MED12S; Mediator complex subunit 12; mediator of RNA polymerase II transcription subunit 12; mediator of RNA polymerase II transcription, subunit 12 homolog; Mopa; OHDOX; OKS; OPA1; OPA-1; OPA-containing protein; OPA-containing protein 1; putative mediator subunit 12; thyroid hormone receptor-associated protein complex 230 kDa component; thyroid hormone receptor-associated protein, 230 kDa subunit; TNRC11; TRAP230; trinucleotide repeat containing 11 (THR-associated protein); trinucleotide repeat containing 11 (THR-associated protein, 230 kDa subunit); trinucleotide repeat-containing gene 11 protein | |
| Med12 | |
| Primary | |
| Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title