missing translation for 'onlineSavingsMsg'
Learn More

MED12 Antibody, Novus Biologicals™

Product Code. 18412131 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412131 25 μL 25µL
18229557 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18412131 Supplier Novus Biologicals Supplier No. NBP18667025ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 1 publication

MED12 Polyclonal specifically detects MED12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:20-1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias ARC240, CAG repeat protein 45, CAGH45trinucleotide repeat containing 11 (THR-associated protein, 230kDa subunit), HOPAFGS1, KIAA0192FG syndrome 1, mediator complex subunit 12OPA-containing protein, OKSsubunit 12 homolog, thyroid hormone receptor-associated protein, 230 kDa subunit, Trap230, TRAP230mediator of RNA polymerase II transcription, subunit 12 homolog (S. cerevisiae), trinucleotide repeat containing 11 (THR-associated protein, 230 kDa subunit), Trinucleotide repeat-containing gene 11 protein
Gene Symbols MED12
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:RLLLYHTHLRPRPRAYYLEPLPLPPEDEEPPAPTLLEPEKKAPEPPKTDKPGAAPPSTEERKKKSTKGKKRSQPATKTEDYGMGPGRSGPYGVTVPPDLLHHPNPGSITHLNYRQGSIGLYTQNQ
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9968
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.