missing translation for 'onlineSavingsMsg'
Learn More

Mecysteine Hydrochloride, TRC

Product Code. 30138718
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138718 5 g 5g
30008405 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138718 Supplier Toronto Research Chemicals Supplier No. TRCM2031005G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18598-63-5
Molecular Formula C4H10ClNO2S
Molecular Weight (g/mol) 171.65
Synonym L-Cysteine Methyl Ester Hydrochloride, (R)-Cysteine Methyl Ester Hydrochloride, Acdrile, Actiol, LJ 48, Methyl (R)-2-Amino-3-mercaptopropanoate Hydrochloride, Methyl L-Cysteinate Hydrochloride, Methyl Cysteine Hydrochloride, Methyl alpha-Amino-beta-mercaptopropionate Hydrochloride, Methyl beta-Mercaptoalanine Hydrochloride, NSC 161611, Pectite, Visclair, Zeotin
IUPAC Name methyl (2R)-2-amino-3-sulfanylpropanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CS

Specifications

InChI Formula InChI=1S/C4H9NO2S.ClH/c1-7-4(6)3(5)2-8;/h3,8H,2,5H2,1H3;1H/t3-;/m0./s1
Quantity 5 g
Formula Weight 171.0121
Shipping Condition Room Temperature
Chemical Name or Material Mecysteine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.