missing translation for 'onlineSavingsMsg'
Learn More

MECR Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18419356 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419356 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419356 Supplier Novus Biologicals Supplier No. NBP281846AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MECR Polyclonal antibody specifically detects MECR in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MECR
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias CGI-63, EC 1.3.1.38, FASN2B, homolog of yeast 2-enoyl thioester reductase, HsNrbf-1, mitochondrial 2-enoyl thioester reductase, mitochondrial trans-2-enoyl-CoA reductase, NBRF1, NRBF-1, NRBF1hsNrbf-1, nuclear receptor binding factor 1, Nuclear receptor-binding factor 1, trans-2-enoyl-CoA reductase, mitochondrial
Host Species Rabbit
Immunogen Rabbit polyclonal MECR antibody was raised against a 15 amino acid peptide near the carboxy terminus of human MECR. The immunogen is located within amino acids 300 - 350 of MECR. Amino Acid Squence: LSQWKKDHSPDQFKE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 51102
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.