Learn More
MECP2 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-10861-100UL
Product Code. 17442473
  • £375.00 / 100µL
Description
The MECP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MECP2. This antibody recognizes human, mouse antigen. The MECP2 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
Specifications
MECP2
Polyclonal
Rabbit
Primary
Human, Mouse
Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite 488
100 μL
4204