missing translation for 'onlineSavingsMsg'
Learn More

Meconin (1.0 mg/mL in Chloroform), TRC

Product Code. 30481940
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481940 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481940 Supplier Toronto Research Chemicals Supplier No. TRCKIT82251X1ML
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 569-31-3
Molecular Formula C10 H10 O4
Quantity 1 mL
Synonym 1(3H)-Isobenzofuranone, 6,7-dimethoxy-, Phthalide, 6,7-dimethoxy- (6CI,7CI,8CI), 6,7-Dimethoxy-1(3H)-isobenzofuranone, 6,7-Dimethoxyphthalide, Meconic lactone, Meconin, Meconine, NSC 35547, Opianyl
InChI Formula InChI=1S/C10H10O4/c1-12-7-4-3-6-5-14-10(11)8(6)9(7)13-2/h3-4H,5H2,1-2H3
SMILES COc1ccc2COC(=O)c2c1OC
IUPAC Name 6,7-dimethoxy-3H-2-benzofuran-1-one
Molecular Weight (g/mol) 194.18
Formula Weight 194.0579
Concentration or Composition (by Analyte or Components) Meconin 1 mg/mL
Chemical Name or Material Meconin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.