missing translation for 'onlineSavingsMsg'
Learn More

Meclofenoxate Hydrochloride, TRC

Product Code. 30137567
Change view
Click to view available options
Quantity:
50 mg
5 g
10 g
Unit Size:
10g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137567 5 g 5g
30135124 10 g 10g
17971065 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137567 Supplier Toronto Research Chemicals Supplier No. TRCM2027705G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3685-84-5
Molecular Formula C12 H16 Cl N O3 . Cl H
Molecular Weight (g/mol) 294.17
Synonym Acetic acid, 2-(4-chlorophenoxy)-, 2-(dimethylamino)ethyl ester, hydrochloride (1:1), Acetic acid, (4-chlorophenoxy)-, 2-(dimethylamino)ethyl ester, hydrochloride (9CI), Acetic acid, (p-chlorophenoxy)-, 2-(dimethylamino)ethyl ester hydrochloride (6CI,8CI), 2-(Dimethylamino)ethyl (p-chlorophenoxy)acetate hydrochloride, Acefen, Acephen, Brenal, Cellative, Centrophenoxine, Centrophenoxine hydrochloride, Cerutil, Dimethylaminoethyl 4-chlorophenoxyacetate hydrochloride, Dimethylaminoethyl p-chlorophenoxyacetate hydrochloride, Helfergin, Lucidril, Lucidryl hydrochloride, Marucotol, Meclofenoxate hydrochloride, Meclophenoxate hydrochloride, Methocynal, Proserout
IUPAC Name 2-(dimethylamino)ethyl 2-(4-chlorophenoxy)acetate;hydrochloride
SMILES Cl.CN(C)CCOC(=O)COc1ccc(Cl)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H16ClNO3.ClH/c1-14(2)7-8-16-12(15)9-17-11-5-3-10(13)4-6-11;/h3-6H,7-9H2,1-2H3;1H
Formula Weight 293.0585
Chemical Name or Material Meclofenoxate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.