missing translation for 'onlineSavingsMsg'
Learn More

Meclofenoxate Hydrochloride 1.0 mg/ml in Acetonitrile (as free base), LoGiCal

Product Code. 30459467
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459467 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459467 Supplier LoGiCal Supplier No. LGCAMP0044.0011
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Meclofenoxate Hydrochloride
CAS 3685-84-5
Analyte or Component Names Meclofenoxate Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C12 H16 Cl N O3 . Cl H
Certifications/Compliance ISO 17034
Synonym Acetic acid, 2-(4-chlorophenoxy)-, 2-(dimethylamino)ethyl ester, hydrochloride (1:1), Acetic acid, (4-chlorophenoxy)-, 2-(dimethylamino)ethyl ester, hydrochloride (9CI), Acetic acid, (p-chlorophenoxy)-, 2-(dimethylamino)ethyl ester hydrochloride (6CI,8CI), 2-(Dimethylamino)ethyl (p-chlorophenoxy)acetate hydrochloride, Acefen, Acephen, Brenal, Cellative, Centrophenoxine, Centrophenoxine hydrochloride, Cerutil, Dimethylaminoethyl 4-chlorophenoxyacetate hydrochloride, Dimethylaminoethyl p-chlorophenoxyacetate hydrochloride, Helfergin, Lucidril, Lucidryl hydrochloride, Marucotol, Meclofenoxate hydrochloride, Meclophenoxate hydrochloride, Methocynal, Proserout
InChI Formula InChI=1S/C12H16ClNO3.ClH/c1-14(2)7-8-16-12(15)9-17-11-5-3-10(13)4-6-11;/h3-6H,7-9H2,1-2H3;1H
Solution Type Single Solution
SMILES Cl.CN(C)CCOC(=O)COc1ccc(Cl)cc1
IUPAC Name 2-(dimethylamino)ethyl 2-(4-chlorophenoxy)acetate;hydrochloride
Solvent or Matrix Acetonitrile
Industry Type Clinical Testing
Molecular Weight (g/mol) 294.17
Quantity 1 mL
Formula Weight 293.0585 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.